Order Entry
Italy
ContactUsLinkComponent
Thermometer, solid stem, similar to ASTM 29 F, white backed, 127,5+132,5:0,1F, capillary tube specially coated inside, with non-we tting blue special liquid, total immersion, max.310x6-8mm, durable pigment, with ice point scale, null
Catalog # AMARA300480-CB
Supplier:  Amarell
Thermometer, solid stem, similar to ASTM 29 F, white backed, 127,5+132,5:0,1F, capillary tube specially coated inside, with non-we tting blue special liquid, total immersion, max.310x6-8mm, durable pigment, with ice point scale, null
Catalog # AMARA300480-CB
Supplier:  Amarell
Frequently Bought Together