Order Entry
Italy
ContactUsLinkComponent
Thermometer similar to BS 1900, SR6/34C, 16+34:0,05°C, length 480mm, total immersion, solid stem, white backed, capillary tube spec ially coated inside, with non-wetting blue special liquid, with ice point scale, null
Catalog # AMARA401930-CB
Supplier:  Amarell
Thermometer similar to BS 1900, SR6/34C, 16+34:0,05°C, length 480mm, total immersion, solid stem, white backed, capillary tube spec ially coated inside, with non-wetting blue special liquid, with ice point scale, null
Catalog # AMARA401930-CB
Supplier:  Amarell
Frequently Bought Together