Order Entry
Italy
Orders LinkContactUsLinkComponent
VWR®, DNA ladders

VWR®, DNA ladders

PCR ladder, 100 − 3000 bp, 100 Tests
Codice catalogo 733-2603
Nome del fornitore:   VWR Chemicals
VWR®, DNA ladders
Codice catalogo 733-2603
Nome del fornitore:  VWR Chemicals
Numero CAS:  
VWR®, DNA ladders
VWR®, DNA ladders

PCR ladder, 100 − 3000 bp, 100 Tests

Numero CAS:  
Price
55,00 €
Log in for availability
1 * 100 Tests
Product Details & Documents

VWR® DNA ladders are convenient dsDNA ladders supplied in a loading buffer, which are ready to use on agarose and SDS DNA gels.

  • Ready to use loading buffer
  • For direct loading and easy visualisation
  • Suitable with TBE and TAE electrophoresis systems
  • Clear, distinct bands
  • Blue dye front running at 100 to 300 bp at 0,5 to 1,5% agarose

Ordering information: Supplied in 0,5 ml packs for 100 lanes.

Specifications

Specifications

  • Pk:
    100 Tests
  • Description:
    PCR ladder, 100 − 3000 bp

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.