Order Entry
Italy
ContactUsLinkComponent
3 vials of 10mL - 22 components 2000mg/l each of Ethylbenzene [CAS:100-41-4] Toluene [CAS:108-88-3] m-Xylene [CAS:108-38-3] o-Xylene [CAS:95-47-6] p-Xylene [CAS:106-42-3] Benzene [CAS:71-43-2] Isopropylbenzene [CAS:98-82-8] Chloroben [ 1 * 3 Ampoul
Catalog # CMPLRD00763410.3
3 vials of 10mL - 22 components 2000mg/l each of Ethylbenzene [CAS:100-41-4] Toluene [CAS:108-88-3] m-Xylene [CAS:108-38-3] o-Xylene [CAS:95-47-6] p-Xylene [CAS:106-42-3] Benzene [CAS:71-43-2] Isopropylbenzene [CAS:98-82-8] Chloroben [ 1 * 3 Ampoul
Catalog # CMPLRD00763410.3