Order Entry
Italy
ContactUsLinkComponent
Detect Rat Neuropilin-1/NRP1 with <20pg/ml sensitivity. Format: 96-well plate with removable strips. 3002190000 CN 0.1 0.045 kg Yes 365 N 20 T2 N/A 1 * 1 KIT
Codice catalogo BSBTEK1343
Supplier:  Boster Bio
undefined
Detect Rat Neuropilin-1/NRP1 with <20pg/ml sensitivity. Format: 96-well plate with removable strips. 3002190000 CN 0.1 0.045 kg Yes 365 N 20 T2 N/A 1 * 1 KIT
Codice catalogo BSBTEK1343
Supplier:  Boster Bio