Order Entry
Italy
Orders LinkContactUsLinkComponent
 

6,8-Dimethyl-4-hydroxyquinoline

Bottle, 1 g
Catalog # APOSOR307846-1G
Supplier:   Apollo Scientific
6,8-Dimethyl-4-hydroxyquinoline
Catalog # APOSOR307846-1G
Supplier:  Apollo Scientific
CAS Number:  203626-58-8
6,8-Dimethyl-4-hydroxyquinoline

Bottle, 1 g

CAS Number:  203626-58-8
Price
291,00 €
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:203626-58-8
  • Formula:C₁₁H₁₁NO
  • MDL Number:MFCD00272458
  • Molecular Weight:173.211 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.