Order Entry
Italy
Orders LinkContactUsLinkComponent
Thioacetamide 98%

Thioacetamide 98%

Bottle, 25 g
Catalog # APOSOR28604-25G
Irritant
Health
Supplier:   Apollo Scientific
Thioacetamide 98%
Catalog # APOSOR28604-25G
Supplier:  Apollo Scientific
CAS Number:  62-55-5
Thioacetamide 98%

Bottle, 25 g

CAS Number:  62-55-5
Price
21,80 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  Ethanethioamide
  • CAS Number:62-55-5
  • Formula:CH₃CSNH₂
  • MDL Number:MFCD00008070
  • Molecular Weight:75.13 g/mol
  • Melting Pt:112…114 °C
  • Storage Temperature:Ambient
  • EINECS:200-541-4
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.