Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Thioacetamide 98%

Bottle, 100 g
Catalog # APOSOR28604-100G
Irritant
Health
Supplier:   Apollo Scientific
Thioacetamide 98%
Catalog # APOSOR28604-100G
Supplier:  Apollo Scientific
CAS Number:  62-55-5
Thioacetamide 98%

Bottle, 100 g

CAS Number:  62-55-5
Price
31,90 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Ethanethioamide
  • CAS Number:62-55-5
  • Formula:CH₃CSNH₂
  • MDL Number:MFCD00008070
  • Molecular Weight:75.13 g/mol
  • Melting Pt:112…114 °C
  • Storage Temperature:Ambient
  • EINECS:200-541-4
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g

Specifications