Order Entry
Italy
Orders LinkContactUsLinkComponent
N-Hydroxysuccinimide 99%

N-Hydroxysuccinimide 99%

Bottle, 100 g
Codice catalogo APOSOR28434-100G
Irritant
Nome del fornitore:   Apollo Scientific
N-Hydroxysuccinimide 99%
Codice catalogo APOSOR28434-100G
Nome del fornitore:  Apollo Scientific
Numero CAS:  6066-82-6
N-Hydroxysuccinimide 99%

Bottle, 100 g

Numero CAS:  6066-82-6
Price
21,80 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  1-Hydroxypyrrolidine-2,5-dione, NHS, HOSu
  • CAS Number:6066-82-6
  • Formula:C₄H₅NO₃
  • MDL Number:MFCD00005516
  • Molecular Weight:115.09 g/mol
  • Melting Pt:94…99 °C
  • Storage Temperature:Ambient
  • EINECS:228-001-3

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.