Order Entry
Italy
Orders LinkContactUsLinkComponent
1-Hydroxy-2-naphthaldehyde

1-Hydroxy-2-naphthaldehyde

Bottle, 5 g
Catalog # APOSOR18562-5G
Irritant
Supplier:   Apollo Scientific
1-Hydroxy-2-naphthaldehyde
Catalog # APOSOR18562-5G
Supplier:  Apollo Scientific
CAS Number:  574-96-9
1-Hydroxy-2-naphthaldehyde

Bottle, 5 g

CAS Number:  574-96-9
Price
151,00 €
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:574-96-9
  • Formula:C₁₁H₈O₂
  • MDL Number:MFCD00092325
  • Molecular Weight:172.1800 g/mol
  • Melting Pt:56 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.