Order Entry
Italy
Orders LinkContactUsLinkComponent
 

1-Hydroxy-2-naphthaldehyde

Bottle, 1 g
Catalog # APOSOR18562-1G
Irritant
Supplier:   Apollo Scientific
1-Hydroxy-2-naphthaldehyde
Catalog # APOSOR18562-1G
Supplier:  Apollo Scientific
CAS Number:  574-96-9
1-Hydroxy-2-naphthaldehyde

Bottle, 1 g

CAS Number:  574-96-9
Price
33,30 €
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:574-96-9
  • Formula:C₁₁H₈O₂
  • MDL Number:MFCD00092325
  • Molecular Weight:172.1800 g/mol
  • Melting Pt:56 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications