Order Entry
Italy
Orders LinkContactUsLinkComponent
2,3-Dibromo-5-picoline

2,3-Dibromo-5-picoline

Bottle, 5 g
Catalog # APOSOR8978-5G
Corrosive
Toxic
Supplier:   Apollo Scientific
2,3-Dibromo-5-picoline
Catalog # APOSOR8978-5G
Supplier:  Apollo Scientific
CAS Number:  29232-39-1
2,3-Dibromo-5-picoline

Bottle, 5 g

CAS Number:  29232-39-1
Price
21,80 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  2,3-Dibromo-5-methylpyridine
  • CAS Number:29232-39-1
  • Formula:C₆H₅Br₂N
  • MDL Number:MFCD04112574
  • Molecular Weight:250.92 g/mol
  • Boiling Pt:270.8 °C
  • Melting Pt:52…56 °C
  • Flash Pt:>110 °C
  • Storage Temperature:Ambient
  • UN:2811
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications