4-Nitro-2-picoline
Bottle, 1 g
Catalog # APOSOR9449-1G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: 2-Methyl-4-nitropyridine
- CAS Number:13508-96-8
- Formula:C₆H₆N₂O₂
- MDL Number:MFCD03095077
- Molecular Weight:138.1200 g/mol
- Melting Pt:42…44 °C
- Storage Temperature:Refrigerator
Specifications
Specifications
- Pack type:Bottle
- Pk:1 g



