Order Entry
Italy
Orders LinkContactUsLinkComponent
2,3-Dichloropropene 97%

2,3-Dichloropropene 97%

Bottle, 1 kg
Codice catalogo APOSOR5736-1KG
Flammable
Irritant
Health
Nome del fornitore:   Apollo Scientific
2,3-Dichloropropene 97%
Codice catalogo APOSOR5736-1KG
Nome del fornitore:  Apollo Scientific
Numero CAS:  78-88-6
2,3-Dichloropropene 97%

Bottle, 1 kg

Numero CAS:  78-88-6
Price
406,00 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  2,3-Dichloro-1-propene
  • CAS Number:78-88-6
  • Formula:C₃H₄Cl₂
  • MDL Number:MFCD00000943
  • Molecular Weight:110.97 g/mol
  • Boiling Pt:94 °C
  • Melting Pt:–83 °C
  • Density:1.209 g/cm³ (25 °C)
  • Flash Pt:10 °C
  • Storage Temperature:Ambient
  • UN:2047
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 kg

Specifications