Order Entry
Italy
Orders LinkContactUsLinkComponent
Selenium dioxide 99,9%

Selenium dioxide 99,9%

Bottle, 500 g
Catalog # APOSIN3175-500G
Supplier:   Apollo Scientific
Selenium dioxide 99,9%
Catalog # APOSIN3175-500G
Supplier:  Apollo Scientific
CAS Number:  7446-08-4
Selenium dioxide 99,9%

Bottle, 500 g

CAS Number:  7446-08-4
Price
999,00 €
Log in for availability
1 * 500 g
Product Details & Documents
  • CAS Number:7446-08-4
  • Formula:SeO₂
  • MDL Number:MFCD00003562
  • Molecular Weight:110.96 g/mol
  • Melting Pt:340…350 °C (subl.)
  • Density:3.95 g/cm³ (25 °C)
  • Storage Temperature:Ambient
  • UN:3283
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.