Order Entry
Italy
Orders LinkContactUsLinkComponent
17-DMAG

17-DMAG

Bottle, 10 mg
Catalog # APOSBID1002-10MG
Irritant
Supplier:   Apollo Scientific
17-DMAG
Catalog # APOSBID1002-10MG
Supplier:  Apollo Scientific
CAS Number:  467214-20-6
17-DMAG

Bottle, 10 mg

CAS Number:  467214-20-6
Price
99,90 €
Log in for availability
1 * 10 mg
Product Details & Documents
Synonyms:  17-Dimethylaminoethylamino-17-demethoxygeldanamycin
  • CAS Number:467214-20-6
  • Formula:C₃₂H₄₈N₄O₈
  • MDL Number:MFCD08457919
  • Molecular Weight:616.8 g/mol
  • Storage Temperature:Freezer
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    10 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.