Order Entry
Italy
Orders LinkContactUsLinkComponent
L(+)-Histidine ≥99%

L(+)-Histidine ≥99%

Bottle, 500 g
Codice catalogo APOSBIH0710-500G
Irritant
Nome del fornitore:   Apollo Scientific
L(+)-Histidine ≥99%
Codice catalogo APOSBIH0710-500G
Nome del fornitore:  Apollo Scientific
Numero CAS:  71-00-1
L(+)-Histidine ≥99%

Bottle, 500 g

Numero CAS:  71-00-1
Price
108,00 €
Log in for availability
1 * 500 g
Product Details & Documents

USP, Pyrogen free

Synonyms:  L-3-Imidazol-4-ylalanine, (S)-(+)-Histidine, (S)-2-Amino-3-(4-imidazolyl)propionic acid, H-His-OH, NSC 137773, L-Histidine
  • CAS Number:71-00-1
  • Formula:C₆H₉N₃O₂
  • MDL Number:MFCD00064315
  • Molecular Weight:155.16 g/mol
  • Melting Pt:282 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:200-745-3

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.