Chromomycin A3
Bottle, 5 mg
Codice catalogo APOSBIC1009-5MG
Nome del fornitore: Apollo Scientific
Chromomycin A3
Codice catalogo APOSBIC1009-5MG
Nome del fornitore: Apollo Scientific
Numero CAS: 7059-24-7
Jump to:
Product Details & Documents
Antibiotic that can serve as a fluorescent DNA stain.
- CAS Number:7059-24-7
- Formula:C₅₇H₈₂O₂₆
- MDL Number:MFCD00043151
- Molecular Weight:1183.30 g/mol
- Melting Pt:182 °C
- Storage Temperature:Freezer
- EINECS:230-348-0
- UN:3462
- ADR:6.1,I
Specifications
Specifications
- Pack type:Bottle
- Pk:5 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



