Order Entry
Italy
Orders LinkContactUsLinkComponent
2-Naphthaldehyde

2-Naphthaldehyde

Bottle, 100 g
Catalog # APOSOR350292-100G
Irritant
Supplier:   Apollo Scientific
2-Naphthaldehyde
Catalog # APOSOR350292-100G
Supplier:  Apollo Scientific
CAS Number:  66-99-9
2-Naphthaldehyde

Bottle, 100 g

CAS Number:  66-99-9
Price
76,70 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  β-Naphthaldehyde
  • CAS Number:66-99-9
  • Formula:C₁₁H₈O
  • MDL Number:MFCD00004094
  • Molecular Weight:156.18 g/mol
  • Boiling Pt:160…19mm °C
  • Melting Pt:58…61 °C
  • Flash Pt:>100 °C
  • Storage Temperature:Freezer
  • EINECS:200-640-2
  • UN:3335
  • ADR:9,III
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.