Order Entry
Italy
Orders LinkContactUsLinkComponent
Amphotericin B

Amphotericin B

Bottle, 1 g
Catalog # APOSBIA0103-1G
Irritant
Supplier:   Apollo Scientific
Amphotericin B
Catalog # APOSBIA0103-1G
Supplier:  Apollo Scientific
CAS Number:  1397-89-3
Amphotericin B

Bottle, 1 g

CAS Number:  1397-89-3
Price
46,40 €
Log in for availability
1 * 1 g
Product Details & Documents

A polyene antifungal antibiotic produced by Streptomyces nodosus.

Synonyms:  (1R,3S,5R,6R,9R,11R,15S,16R,17R,18S,19E,21E,23E,25E,27E,29E,31E,33R,35S,36R,37S)-33-(((2R,3S,4S,5S,6R)-4-Amino-3,5-dihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-1,3,5,6,9,11,17,37-octahydroxy-15,16,18-trimethyl-13-oxo-14,39-dioxabicyclo[33.3.1]nonatriaconta-19,21,23,25,27,29,31-heptaene-36-carboxylic acid
  • CAS Number:1397-89-3
  • Formula:C₄₇H₇₃NO₁₇
  • MDL Number:MFCD00877763
  • Molecular Weight:924.08 g/mol
  • Storage Temperature:Refrigerator

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications