Order Entry
Italy
Orders LinkContactUsLinkComponent
(±)-1,2-Octanediol

(±)-1,2-Octanediol

Bottle, 2,5 kg
Catalog # APOSBIB6291-2.5KG
Irritant
Supplier:   Apollo Scientific
(±)-1,2-Octanediol
Catalog # APOSBIB6291-2.5KG
Supplier:  Apollo Scientific
CAS Number:  1117-86-8
(±)-1,2-Octanediol

Bottle, 2,5 kg

CAS Number:  1117-86-8
Price
514,00 €
Log in for availability
1 * 2,5 kg
Product Details & Documents
Synonyms:  1,2-Octanediol, (±)-1,2-Octylene glycol, (±)-1,2-Dihydroxy octane
  • CAS Number:1117-86-8
  • Formula:C₈H₁₈O₂
  • MDL Number:MFCD00010738
  • Molecular Weight:146.23 g/mol
  • Boiling Pt:131…132 °C
  • Melting Pt:28…31 °C
  • Flash Pt:138 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    2,5 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.