Order Entry
Italy
Orders LinkContactUsLinkComponent
(±)-1,2-Octanediol

(±)-1,2-Octanediol

Bottle, 1 kg
Catalog # APOSBIB6291-1KG
Irritant
Supplier:   Apollo Scientific
(±)-1,2-Octanediol
Catalog # APOSBIB6291-1KG
Supplier:  Apollo Scientific
CAS Number:  1117-86-8
(±)-1,2-Octanediol

Bottle, 1 kg

CAS Number:  1117-86-8
Price
268,00 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  1,2-Octanediol, (±)-1,2-Octylene glycol, (±)-1,2-Dihydroxy octane
  • CAS Number:1117-86-8
  • Formula:C₈H₁₈O₂
  • MDL Number:MFCD00010738
  • Molecular Weight:146.23 g/mol
  • Boiling Pt:131…132 °C
  • Melting Pt:28…31 °C
  • Flash Pt:138 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 kg

Specifications