Xylan (from beechwood)
Bottle, 25 g
Nome del fornitore: Apollo Scientific
Xylan (from beechwood)
Codice catalogo APOSBI3856-25G
Nome del fornitore: Apollo Scientific
Numero CAS: 9014-63-5
Jump to:
Product Details & Documents
- CAS Number:9014-63-5
- MDL Number:MFCD00082148
- Melting Pt:300 °C
- Storage Temperature:Ambient
- EINECS:232-760-6
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Bottle
- Pk:25 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



