Order Entry
Italy
Orders LinkContactUsLinkComponent
Xylan (from beechwood)

Xylan (from beechwood)

Bottle, 100 g
Catalog # APOSBI3856-100G
Irritant
Supplier:   Apollo Scientific
Xylan (from beechwood)
Catalog # APOSBI3856-100G
Supplier:  Apollo Scientific
CAS Number:  9014-63-5
Xylan (from beechwood)

Bottle, 100 g

CAS Number:  9014-63-5
Price
283,00 €
Log in for availability
1 * 100 g
Product Details & Documents
  • CAS Number:9014-63-5
  • MDL Number:MFCD00082148
  • Melting Pt:300 °C
  • Storage Temperature:Ambient
  • EINECS:232-760-6

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g

Specifications