Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Columns for large biomolecule analysis, CIMmultus™

Column with 5/16-24 UNF coned port, 1/8" OD tubing connection, Preparative, QA (quaternary amine), 2 µm, 8 ml, Polymer monolith
Codice catalogo HISM411.5113-2
Nome del fornitore:   HICHROM
Columns for large biomolecule analysis, CIMmultus™
Codice catalogo HISM411.5113-2
Nome del fornitore:  HICHROM
Columns for large biomolecule analysis, CIMmultus™

Column with 5/16-24 UNF coned port, 1/8" OD tubing connection, Preparative, QA (quaternary amine), 2 µm, 8 ml, Polymer monolith

Price
Inquire For Price
Log in for availability
1 * 1 pezzi
Product Details & Documents

CIMmultus™ columns are based on an innovative monolithic technology. Distinguished by large pores/flow through channels these columns are designed for the production, purification, and analytics of large proteins (IgG, IgM, PEGylated proteins), viruses (AAV, adenovirus, lentivirus, influenza, bacteriophages), VLPs, exosomes and pDNA for example.

  • Rapid, high resolution analysis
  • Flow independent separation
  • High sensitivity and reproducibility
  • Operation at high volumetric flow rates
  • Larger scale columns available and in cGMP compliant formats

Distinguished by their flow independent resolution, these columns offer high efficiency, high speed, and high yield in downstream processing for large biomolecules. Due to their performance properties, CIMmultus™ columns can be used as single-use disposable or multi-use columns.

Certifications: USP Class VI.

Caution: Do not expose the column housing to pure acetone.

Specifications
  • Column Format:
    Preparative
  • Particle Size:
    2 µm
  • Capacity:
    8 ml
  • Description:
    Column with 5/16-24 UNF coned port, 1/8" OD tubing connection
  • Seq:
    30
  • Phase:
    QA (quaternary amine)
  • Packing material:
    Polymer monolith