Order Entry
Italy
ContactUsLinkComponent
LipidSpot™ 488, 1000X in DMSO, rapidly stains intracellular lipid droplets with bright green fluorescence in live or fixed cells, wi th no wash step required. LipidSpot™ 488 shows minimal background staining of cellular membranes or other organelles, 1 * 20 µl
Catalog # 70065-T
Supplier:  Biotium
LipidSpot™ 488, 1000X in DMSO, rapidly stains intracellular lipid droplets with bright green fluorescence in live or fixed cells, wi th no wash step required. LipidSpot™ 488 shows minimal background staining of cellular membranes or other organelles, 1 * 20 µl
Catalog # 70065-T
Supplier:  Biotium
Frequently Bought Together