0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Italy
ContactUsLinkComponent
LINOLENIC ACID METHYL ESTER 1 * 100 mg
EHERC14635900
:
EHRENSTORFER
LINOLENIC ACID METHYL ESTER 1 * 100 mg
EHERC14635900
:
EHRENSTORFER
Frequently Bought Together
Supelco® Cerilliant® Organic Reference Standard, 17β-Estradiol-2,3,4-¹³C₃ solution
: Merck
Indium Standard for ICP 1 * 100 mL
: Merck
Benzotriazole
: Hach
Olopatadine Related Compounds, Reference Standard
: USP
PCB-Mix 4 10 ng/µl 1 * 10 mL
: EHRENSTORFER
CLOZAPINE PEAK IDENTIFICATION 1 * 0,41 mg
: EDQM
Tolfenpyrad certified 1 * 0,1 g
: EHRENSTORFER
Mechlorethamine Hydrochloride, Reference Standard
: USP
VISCOSITY STANDARD, GENERAL PURPOSE, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 20.4 MPA.S (25 C) 1 * 500 mL
: PARAGON SCIENTIFIC
Triphenyl phosphate solution, NMR reference standard, Supelco®
: Merck
Pharmaceutical secondary standard, tolterodine tartrate, (CRM), Supelco®
: Merck
WFI/Acetone Solution (99.75%/0.25%) 500ml in a HDPE mouser bottle is 0.62KG 1 * 500 mL
: REAGECON
Ampicillin, Reference Standards
: USP
Farnesol (mixture of isomers)
: EHRENSTORFER