Order Entry
Italy
Orders LinkContactUsLinkComponent
Accessories for Automatic Capillary Viscometers, ViscoSystem® AVC 370
Accessories for Automatic Capillary Viscometers, ViscoSystem® AVC 370
VZ 8527 PTFE hose set for high-aggressive media, contains: VZ 8542, VZ 8543, VZ 7012, VZ 7211, VZ 7212, 2×VZ 7215
Catalog # SCOI285415885
Supplier:   SI Analytics
Accessories for Automatic Capillary Viscometers, ViscoSystem® AVC 370
Catalog # SCOI285415885
Supplier:  SI Analytics
Accessories for Automatic Capillary Viscometers, ViscoSystem® AVC 370
Accessories for Automatic Capillary Viscometers, ViscoSystem® AVC 370
VZ 8527 PTFE hose set for high-aggressive media, contains: VZ 8542, VZ 8543, VZ 7012, VZ 7211, VZ 7212, 2×VZ 7215
Price
1 260,00 €
Log in for availability
1 * 1 pezzi
Specifications
  • Description:
    VZ 8527 PTFE hose set for high-aggressive media, contains: VZ 8542, VZ 8543, VZ 7012, VZ 7211, VZ 7212, 2×VZ 7215
  • Seq:
    52