Order Entry
Italy
ContactUsLinkComponent
pH combination electrode with Memosens® screw plug head (ATEX II 1/G),NTC 30K, low temperature version, glass shaft, SILAMID® ref., ceramic diaphr., press. RheoLid ref. system, sphere membrane, A glass, Memosens® screw plug head, length 225 mm 1 * 1 items
Catalog # SCOI285114110
Supplier:  SI Analytics
pH combination electrode with Memosens® screw plug head (ATEX II 1/G),NTC 30K, low temperature version, glass shaft, SILAMID® ref., ceramic diaphr., press. RheoLid ref. system, sphere membrane, A glass, Memosens® screw plug head, length 225 mm 1 * 1 items
Catalog # SCOI285114110
Supplier:  SI Analytics