Order Entry
Italy
ContactUsLinkComponent
Easy Purchase Pack AQ brand, 1ml 8x40mm amber borosilicate glass autosampler vials for Waters 96 position autosamplers. Includes pol yethylene caps with penetration point. 250/pack. 1 * 100 items
Codice catalogo MICS95001-0AP
Supplier:  MICROSOLV
undefined
Easy Purchase Pack AQ brand, 1ml 8x40mm amber borosilicate glass autosampler vials for Waters 96 position autosamplers. Includes pol yethylene caps with penetration point. 250/pack. 1 * 100 items
Codice catalogo MICS95001-0AP
Supplier:  MICROSOLV
Frequently Bought Together