Spill containment caddy, EcoPolyBlend™
Spill containment caddy, yellow, indoor/outdoor, 250 l sump capacity, load capacity 567 kg, 57 kg, 813×1829×660 mm
Catalog # 119-0385
Supplier: JUSTRITE MANUFACTURING
Jump to:
Product Details & Documents
Made up of 90% recycled polyethylene.
- Single-piece, double-walled seamless design eliminates leaks
- Ergonomic design with tilt/stop feature minimises back strain
- Rubber, 254 mm wheels roll over bumpy terrain use indoors or out
They offer convenient handling and dispensing of hazardous chemicals with complete assurance of compliance with EPA and SPCC regulations. It is the ideal choice for managing either 110 or 200 litre drums and the nylon strap holds drum securely in place.
Specifications
- Weight:57 kg
- Description:Spill containment caddy, yellow, indoor/outdoor, 250 l sump capacity, load capacity 567 kg
- Seq:01
- W×D×H:813×1829×660 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



