Chloroquine diphosphate salt 98%
Glass bottle, 100 g
Codice catalogo ACRO455241000
Nome del fornitore: Thermo Fisher Scientific
Chloroquine diphosphate salt 98%
Codice catalogo ACRO455241000
Nome del fornitore: Thermo Fisher Scientific
Numero CAS: 50-63-5
Jump to:
Product Details & Documents
Synonyms: N4-(7-Chloroquinolin-4-yl)-N1,N1-diethylpentane-1,4-diamine bis(phosphate), N4-(7-Chloro-4-quinolyl)-N1,N1-diethyl-1,4-pentanediamine Diphosphate, 7-Chloro-4-[4-(diethylamino)-1-methylbutylamino]quinoline Diphosphate, Chloroquine phosphate
- CAS Number:50-63-5
- Formula:C₁₈H₂₆ClN₃
- MDL Number:MFCD00069852
- Molecular Weight:515.86 g/mol
- Melting Pt:200 °C
- Storage Temperature:Ambient
Specifications
Specification Test Results
- Appearance:White to beige Powder
- Infrared spectrum:Conforms
- Loss on drying:≤2 % (105°C, 16 h)
- HPLC:≥97.5 %
Specifications
- Pack type:Glass bottle
- Pk:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



