Order Entry
Italy
Orders LinkContactUsLinkComponent
Ribbons for label printer BBP™11

Ribbons for label printer BBP™11

Thermal transfer printer ribbon, halogen free, 6000 series, width 110 mm, BBP™11, 70 m, Black
Catalog # BRDY196516
Supplier:   Brady
Ribbons for label printer BBP™11
Catalog # BRDY196516
Supplier:  Brady
Ribbons for label printer BBP™11
Ribbons for label printer BBP™11

Thermal transfer printer ribbon, halogen free, 6000 series, width 110 mm, BBP™11, 70 m, Black

Price
50,70 €
Log in for availability
1 * 1 Roll
Product Details & Documents
Specifications
  • Colour:
    Black
  • Description:
    Thermal transfer printer ribbon, halogen free, 6000 series, width 110 mm
  • Length:
    70 m
  • Seq:
    16
  • For:
    BBP™11
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.