Order Entry
Italy
Orders LinkContactUsLinkComponent
 

CF® Dye dCTP conjugates

CF®568-dCTP, lyophilised, 5 Nmo
  40055-T
 :   Biotium
CF® Dye dCTP conjugates
  40055-T
 :  Biotium
 :  
CF® Dye dCTP conjugates

CF®568-dCTP, lyophilised, 5 Nmo

 :  
 
166,00 €
   
1 * 5 Nmo
 

CF® dye-dCTP can be used to synthesise probes for in situ hybridisation, microarray, or to synthesise labelled DNA probes for in situ hybridisation and nucleic acid blotting applications.

  • Synthesise exceptionally bright and photostable fluorescent DNA probes
  • Detect by fluorescence microscopy, flow cytometry, or in gel
  • Compatible with fixed cells or tissue sections
  • Choice of nine CF® dye colours from green to far-red
  • CF® dyes have superior brightness, photostability, and are water-soluble

 : CF™594 dCTP is not compatible with PCR labelling of DNA probes.

 

 

  • Pk:
    5 Nmo
  • Description:
    CF®568-dCTP, lyophilised
  • Cat. No.:
    40055-T
  • Excitation max.:
    562 nm
  • Emission max.:
    583 nm

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.