Order Entry
Italy
ContactUsLinkComponent
Vane probe dia 16mm, including non-rotatable telescope with scaling to max. 960mm, with integrated measurement button, for velocity measurements in ducts measuring range : +0.6 +50 m/s -10 +70°c 1 * 1 items
Catalog # TEST0635.9542
Supplier:  TESTO
Vane probe dia 16mm, including non-rotatable telescope with scaling to max. 960mm, with integrated measurement button, for velocity measurements in ducts measuring range : +0.6 +50 m/s -10 +70°c 1 * 1 items
Catalog # TEST0635.9542
Supplier:  TESTO
Frequently Bought Together