Order Entry
Italy
ContactUsLinkComponent
individual inhibitor specific for papain and other cystein proteases like cathepsin b&l. individual protease inhibitors are proven t o inhibit > 95% of all proteases. 1 * 25 mg
Catalog # GENO786-985
Supplier:  G-Biosciences
individual inhibitor specific for papain and other cystein proteases like cathepsin b&l. individual protease inhibitors are proven t o inhibit > 95% of all proteases. 1 * 25 mg
Catalog # GENO786-985
Supplier:  G-Biosciences
Frequently Bought Together