Order Entry
Italy
Orders LinkContactUsLinkComponent
Potassium hydroxide solution 2N

Potassium hydroxide solution 2N

Catalog # GENO786-929
Supplier:   G-Biosciences
Potassium hydroxide solution 2N
Catalog # GENO786-929
Supplier:  G-Biosciences
CAS Number:  1310-58-3
Potassium hydroxide solution 2N
CAS Number:  1310-58-3
Price
172,00 €
Log in for availability
1 * 500 mL
Product Details & Documents

A strong, standardized alkaline solution commonly used in biochemistry for pH adjustment, tissue digestion, protein hydrolysis, and cell lysis.

  • Synonym: Caustic potash CAS number: 1310-58-3 Molecular formula: KOH Molecular weight: 56.11

Its strong base properties make it essential in various analytical and preparative procedures where precise control of alkalinity is required.

Synonyms:  Potash caustic, Caustic potash
  • CAS Number:1310-58-3
  • Formula:KOH
  • MDL Number:MFCD00003553
  • Molecular Weight:56.11 g/mol

Documents