Order Entry
Italy
ContactUsLinkComponent
a ready-to-use three-color protein standard with 13 prestained proteins covering a wide range of molecular weights:5-245kda in tris -glycine buffer or3.5-235kda in bis-tris (mops) buffer orbis-tris (mes) buffer).proteins are covalently coupled wi 1 * 100 items
Catalog # GENO786-419
Nome del fornitore:  G-Biosciences
a ready-to-use three-color protein standard with 13 prestained proteins covering a wide range of molecular weights:5-245kda in tris -glycine buffer or3.5-235kda in bis-tris (mops) buffer orbis-tris (mes) buffer).proteins are covalently coupled wi 1 * 100 items
Catalog # GENO786-419
Nome del fornitore:  G-Biosciences
Informazioni correlate
Frequently Bought Together