Order Entry
Italy
Orders LinkContactUsLinkComponent
Combined density and refractometry standards

Combined density and refractometry standards

1-bromonaphthalene, 9 ml, 1,658, 1
Catalog # METT51338014
Supplier:   Mettler - Toledo
Combined density and refractometry standards
Catalog # METT51338014
Supplier:  Mettler - Toledo
CAS Number:  
Combined density and refractometry standards

1-bromonaphthalene, 9 ml, 1,658, 1

CAS Number:  
Price
269,00 €
Log in for availability
1 * 1 pezzi
Product Details & Documents

These combined reference standards are intended to reduce uncertainty, eliminate time-consuming and error prone sample transfer and facilitate fully automatic performance verification.

Documents

Specifications

Specifications

  • Pk:
    1
  • Description:
    1-bromonaphthalene, 9 ml
  • Refractive index:
    1,658

Specifications