Combined density and refractometry standards
2,4-dichlorotoluene, 9 ml, 1,546, 1
Supplier: Mettler - Toledo
Combined density and refractometry standards
Catalog # METT51338013
Supplier: Mettler - Toledo
CAS Number:
Jump to:
Product Details & Documents
These combined reference standards are intended to reduce uncertainty, eliminate time-consuming and error prone sample transfer and facilitate fully automatic performance verification.
Certificates
Search by Lot NumberSpecifications
Specifications
- Pk:1
- Description:2,4-dichlorotoluene, 9 ml
- Refractive index:1,546



