Flask for collecting nitrogen 2l, with cylindrical needle valve,300x100mm, st29/32,acc. to schlenk, null
WITG0.778.104.V
Flask for collecting nitrogen 2l, with cylindrical needle valve,300x100mm, st29/32,acc. to schlenk, null
WITG0.778.104.V
Frequently Bought Together
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection