Order Entry
Italy
ContactUsLinkComponent
Flask for collecting nitrogen 1l, 300x80mm, st14/23,acc. to schlenk, null
Catalog # WITG0.778.008
Supplier:  witeg Labortechnik
Flask for collecting nitrogen 1l, 300x80mm, st14/23,acc. to schlenk, null
Catalog # WITG0.778.008
Supplier:  witeg Labortechnik
Frequently Bought Together