Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Funnels, powder

100 mm, 25 mm, 30 mm
  KAVA632413103100
 :   Kavalier
Funnels, powder
  KAVA632413103100
 :  Kavalier
Funnels, powder
Funnels, powder

100 mm, 25 mm, 30 mm

 
2 760,00 €
   
1 * 10 pezzi
 

Borosilicate glass 3,3.

  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes

With smooth internal walls. For transferring powdery materials. Also suitable for use with liquids. Funnel rim is ground and fire-polished.

 
  • Funnel O-Ø:
    100 mm
  • Seq:
    06
  • Stem O-Ø:
    25 mm
  • Stem length:
    30 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.