This fluorescein tyramide conjugate is used for tyramide signal amplification (TSA)1,2,3, which is an enzyme-mediated detection meth od that uses horseradish peroxidase (HRP) to generate high-density labeling of a target protein or nucleic acid in si 1 * 0,5 mg
BTIU96018
: Biotium
Frequently Bought Together