Capsule Quartz glass in application temperatures of up to 1050°C (maximum 1600°C), circular, low form, with lid, translucent, hig h chemical resistance and purity. Diameter 54mm, height 12mm. The durability or operating cycles of our Capsule Quartz g 1 * 1 items
Catalog # TSLA73031
Supplier: TECHNICAL QUARTZ SOLUTIONS
Frequently Bought Together