Capsule Quartz glass in application temperatures of up to 1050°C (maximum 1600°C), circular, low form, translucent, high chemical resistance and purity. Diameter 53mm, height 13mm. The durability or operating cycles of our Capsule Quartz glass s mai 1 * 1 items
Catalog # TSLA73028
Supplier: TECHNICAL QUARTZ SOLUTIONS
Frequently Bought Together