MEDIUM ACETAMIDE BROTH 1 * 500 g
Catalog # HIMEM148-500G
Supplier: HIMEDIA
Frequently Bought Together
Ethanol denatured ≥96%, meets analytical specification of Ph.Eur. (denaturated with 1%iPr-OH,1% MEK,0.001% Bitrex®)
Supplier: Thermo Fisher Scientific
2-Butoxyethanol 99%, extra pure
Supplier: Thermo Fisher Scientific
Cyclohexane ≥99%
Supplier: Thermo Fisher Scientific
11 components: Ca 2mg/l Zr 5mg/l K 5mg/l Zn 5mg/l Na 5mg/l Fe 5mg/l Al 5mg/l Ti 5mg/l Si 5mg/l P 10mg/l Mn 5mg/l in HNO3 5% 1 * 1 L
Supplier: CUSTOM MADE CHEMICALS LAB
p-Xylene ≥99.0% (by GC)
Supplier: TCI
Chloroform ≥99.5% stabilised
Supplier: Honeywell Chemicals
Ethanol absolute Ph. Eur., J.T.Baker®
Supplier: Avantor
Ethyl acetate ≥99.5% for HPLC
Supplier: Thermo Fisher Scientific
N,N-Dimethylacetamide, extra dry 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific
2,2,4-Trimethylpentane ≥99%, Reagent Grade
Supplier: Honeywell Chemicals
Dichloromethane 99.8% stabilised for HPLC
Supplier: Thermo Fisher Scientific
Dehydrated Media
Supplier: THERMO FISHER DIAGNOSTICS
4,4'-Bis(dimethylamino)diphenylmethane 98%
Supplier: Thermo Fisher Scientific