The proteasome is a multicatalytic proteinase complex with a highly ordered ring-shaped 20S core structure. The core structure is co mposed of 4 rings of 28 non-identical subunits 2 rings are composed of 7 alpha subunits and 2 rings are composed of 1 * 100 µG
Catalog # BWRLBS8845
Supplier: Bioworld Technology
Frequently Bought Together