Order Entry
Italy
ContactUsLinkComponent
The 5-AMP activated protein kinase (AMPK), a member of the SNF1 (sucrose non fermentor) kinase family, is a heterotrimeric protein c omprise of alpha (63 kDa), beta (30 kDa) and gamma (38 kDa) subunits. 1 * 100 µG
Catalog # BWRLBS8823
The 5-AMP activated protein kinase (AMPK), a member of the SNF1 (sucrose non fermentor) kinase family, is a heterotrimeric protein c omprise of alpha (63 kDa), beta (30 kDa) and gamma (38 kDa) subunits. 1 * 100 µG
Catalog # BWRLBS8823
Frequently Bought Together