Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these su bunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. 1 * 100 µG
Catalog # BWRLBS8507
Supplier: Bioworld Technology
Frequently Bought Together